
2-Phenylpropionic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:492-37-5
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00002650
:150.18 g/mol
Boiling Pt:267°C
Melting Pt:–5 °C
Density:1,1 g/cm³ (20 °C)
Flash Pt:148°C
EINECS:207-752-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together