L(+)-Ascorbic acid ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:50-81-7
:
Formula:C₆H₈O₆
MDL Number:MFCD00064328
:176.13 g/mol
Merck Index:14,00830
Boiling Pt:553 °C (1013 hPa)
Melting Pt:192°C
Density:1,694 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-066-2
REACH:exempt
Specification Test Results
- Appearance:White powder to crystal
- Purity (Iodometric Titration):min. 99.0 %
- Specific rotation [a]20/D:+20.5 to +21.5deg (C=10, H2O)
- Solubility in Water:almost transparency