
2-Benzoylpyridine ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:91-02-1
:
Formula:C₁₂H₉NO
MDL Number:MFCD00006300
:183.21 g/mol
Boiling Pt:317 °C (1013 hPa)
Melting Pt:40…43 °C
Density:1,171 g/cm³ (0 °C)
EINECS:202-034-3
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 99.0 %
- Melting point:42.0 to 46.0 deg-C
- Solubility in Methanol:almost transparency