


1-Ethyl-2-pyrrolidone ≥98.0% (by GC)
: TCI
:2687-91-4
:
:C₆H₁₁NO
:MFCD00003199
:113.16 g/mol
:218°C
:0,9979 g/cm³ (20 °C)
:76°C
:220-250-6
:GHS05
:GHS07
:GHS08
:Ambient
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %