2-Furaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:98-01-1
:
Formula:C₅H₄O₂
MDL Number:MFCD00003229
:96.09 g/mol
Merck Index:14,04304
Boiling Pt:160°C
Melting Pt:–37°C
Density:1,15 g/cm³ (20 °C)
Flash Pt:59°C
Storage Temperature:Ambient
EINECS:202-627-7
UN:1199
ADR:6.1,II
:GHS02
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %