2-Bromobenzyl alcohol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:18982-54-2
:
Formula:C₇H₇BrO
MDL Number:MFCD00004600
:187.04 g/mol
Boiling Pt:255 °C (1013 hPa)
Melting Pt:80…82 °C
EINECS:242-719-4
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:79.0 to 82.0 deg-C