
2-Bromo-m-toluic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:53663-39-1
:
Formula:C₈H₇BrO₂
MDL Number:MFCD00079721
:215.05 g/mol
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:135.0 to 139.0 deg-C