
Tetraphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2751-90-8
:
Formula:C₂₄H₂₀BrP
MDL Number:MFCD00011915
Melting Pt:299…300 °C
:419.3 g/mol
EINECS:220-393-4
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in hot Water:almost transparency