Funnels with short stem
Supplier: GLASWARENFABRIK KARL HECHT
Certificates
About this item
Funnels, made of HDPE, translucent. With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses.
- Good chemical resistance
- Suspension eye for easy storage
- Easy to clean