Vacuum butterfly valves, VS series
The VS, VS C and VS B series valves are butterfly-type valves which offers very good conductance. The VS C series features a valve seal made of perfluorelastomer material (FFKM).
- Compact, economic design
- High conductance, low leakage rate
- Gas-tight against atmosphere in both positions
- Helium leak-tested
- Easy to grip, lever-operated, with position indicator