
1,1,3,3-Tetramethylbutyl isocyanide for synthesis, Sigma-Aldrich®
8.18105.0005
: Merck
:
- Pack type:Glass bottle
- Pk:5 ml
- Seq:2210
- Solubility:(20 °C) insoluble
- Molar Mass:139.24 g/mol
- Boiling Point:55 - 56 °C (15 hPa)
- Flash Point:51 °C
- Refractive Index:1.4178 (30 °C, 589 nm)
- Density:0.80 g/cm3 (20 °C)
- Synonyms:Thiocarbamide, 1,1,3,3,-Tetramethylbutylisonitrile, Walborsky reagent, tert-Octylisonitrile
- Hill Formula:C9H17N
- Chemical Formula:(CH3)3CCH2C(CH3)2NC
:
:139.2380 g/mol
:6.1 (3),II
:M-Clarity™ quality level = MQ200
:238-577-8
:3275
:GHS02
:Ambient
:14542-93-9
:C₉H₁₇N
:MFCD00000003