
Bis(pentamethylcyclopentadienyl)titanium(IV) dichloride
Catalog # 39570.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:389.225 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
CAS Number:11136-36-0
Formula:C₂₀H₃₀Cl₂Ti
MDL Number:MFCD00049153