Labelling tapes, industrial grade vinyl tape
Supplier: MICRONOVA
Certificates
About this item
Processed in a controlled environment, tapes are wound on a plastic core and single bagged. Ideal for wheel taping and colour coding applications such as spray bottle identification.
- Forms strong bond without leaving adhesive residue
- Can be written on with cleanroom pen
- Different colours available
Length: 32,9 m