Luminol (3-Aminophthalhydrazide)
Catalog # MOLEM19611311
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:177.16 g/mol
EINECS:208-309-4
CAS Number:521-31-3
Formula:C₈H₇N₃O₂
MDL Number:MFCD00006890