Diphenylboric acid-2-aminoethyl ester (2-APB) ≥98.0% for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:524-95-8
:
Formula:C₁₄H₁₆BNO
MDL Number:MFCD00014823
:225.1 g/mol
:M-Clarity™ quality level = MQ200
EINECS:208-366-5
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- CAS:524-95-8
- Synonyms:Diphenylboric acid-2-aminoethyl ester
- Hill Formula:C₁₄H₁₆BNO
- Molar Mass:225.09 g/mol
- Melting Point:189 - 194 °C
- Storage:Store below +15°C.
- Assay (HClO₄):≥ 98,0 %(m)
- Identity (IR):conforms