Order Entry
Finland
ContactUsLinkComponent
 
Diphenylboric acid-2-aminoethyl ester (2-APB) 98%
 
undefined
Diphenylboric acid-2-aminoethyl ester (2-APB) 98%
 

 

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    190 to 194 °C
  • Titration with HClO4:
    ≥97.5 %