Order Entry
Finland
ContactUsLinkComponent
Nalidixic acid sodium salt
Nalidixic acid sodium salt
Catalog # MOLE75525039-25G
Supplier:  Molekula
CAS Number:  
Nalidixic acid sodium salt
Catalog # MOLE75525039-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :254.22 g/mol
EINECS:222-159-7
CAS Number:3374-05-8
Formula:C₁₂H₁₁N₂NaO₃
MDL Number:MFCD00064376