Order Entry
Finland
Orders LinkContactUsLinkComponent
Nalidixic acid

Nalidixic acid

Glass bottle for solids, 25 g
Catalog # MOLEM18439941
Health
Irritant
Marketsource
Supplier:   Molekula
Nalidixic acid
Catalog # MOLEM18439941
Supplier:  Molekula
Marketsource
CAS Number:  389-08-2
Nalidixic acid

Glass bottle for solids, 25 g

CAS Number:  389-08-2
Price
125,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:389-08-2
  • Formula:C₁₂H₁₂N₂O₃
  • MDL Number:MFCD00006884
  • Molecular Weight:232.24 g/mol
  • Storage Temperature:Ambient
  • EINECS:206-864-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications