Order Entry
Finland
Orders LinkContactUsLinkComponent
Nalidixic acid

Nalidixic acid

Glass bottle for solids, 100 g
Catalog # A1894.0100
Health
Irritant
Supplier:   PanReac AppliChem
Nalidixic acid
Catalog # A1894.0100
Supplier:  PanReac AppliChem
CAS Number:  389-08-2
Nalidixic acid

Glass bottle for solids, 100 g

CAS Number:  389-08-2
Price
854,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:389-08-2
  • Formula:C₁₂H₁₂N₂O₃
  • MDL Number:MFCD00006884
  • Molecular Weight:232.24 g/mol
  • Storage Temperature:Ambient
  • EINECS:206-864-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.