Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rosolic acid (Aurin)
Rosolic acid (Aurin)
Rosolic acid (Aurin)
Rosolic acid (Aurin)
Supplier:
CHROMA
Certificates
Expand image
Rosolic acid (Aurin)
Supplier:
CHROMA
About this item
CAS Number:
603-45-2
:
Formula:
C₁₉H₁₄O₃
MDL Number:
MFCD00001624
:
290.32 g/mol
EINECS:
210-041-8