


1,2-Diphenylhydrazine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:122-66-7
:
Formula:C₁₂H₁₂N₂
MDL Number:MFCD00003012
Storage Temperature:Refrigerator
EINECS:204-563-5
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:125.0 to 128.0 deg-C
- Solubility in Methanol:almost transparency