
Allylboronic acid pinacol ester ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:72824-04-5
:
Formula:C₉H₁₇BO₂
MDL Number:MFCD00013347
:168.05 g/mol
Boiling Pt:50…53 °C (5 mmHg)
Density:0.890
Flash Pt:46 °C (140 °F)
Storage Temperature:Ambient
UN:3272
ADR:3,III
:GHS02