Acetoxyacetyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13831-31-7
:
Formula:C₄H₅ClO₃
MDL Number:MFCD00011535
:136.53 g/mol
Boiling Pt:55 °C (12 mmHg)
Density:1.270
Flash Pt:71 °C (160 °F)
Storage Temperature:Ambient
EINECS:237-542-4
UN:3265
ADR:8,II
:GHS05
:GHS07