Order Entry
Finland
ContactUsLinkComponent
Alcian blue 8GX, powder, Sigma-Aldrich®
Alcian blue 8GX, powder, Sigma-Aldrich®
Certificates
undefined
Alcian blue 8GX, powder, Sigma-Aldrich®
Certificates

Specification Test Results

  • Appearance (Colour):
    Dark Blue to Very Dark Blue
  • Appearance (Form):
    Powder
  • Wavelength:
    615 - 630 nm
  • E1%
    615-630 nm in H₂O:
    ≥190
  • Dye Content
    UV Spectroscopy, Based on E1% = 425 at 615-630 nm:
    45 - 65 %
  • Solubility (Colour) 1 mg/ml, H₂O:
    Dark Blue to Very Dark Blue and Dark Blue-Green to Very Dark Blue-Green

Specification Test Results

Related Information