Thionin(acetate), pure, high purity biological stain
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78338-22-4
:
Formula:C₁₄H₁₃N₃O₂S
MDL Number:MFCD00081194
:287.34 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark green Crystalline powder or crystals
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Dye content:≥85 %
- UV Lambda max:597 to 601 nm (in water)