Order Entry
Finland
ContactUsLinkComponent
7-(Diethylamino)-4-methylcoumarin 99%
7-(Diethylamino)-4-methylcoumarin 99%
Certificates
undefined
7-(Diethylamino)-4-methylcoumarin 99%

Specification Test Results

  • Appearance:
    Light beige to light purple Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    70 to 73 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information