

Diethyl (cyanomethyl)phosphonate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2537-48-6
:
Formula:C₆H₁₂NO₃P
MDL Number:MFCD00001893
:177.14 g/mol
Boiling Pt:132…135 °C (1.2 mmHg)
Density:1.094
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:219-806-0
UN:3278
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together