Order Entry
Finland
ContactUsLinkComponent
(Bromomethyl)triphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
Irritant
(Bromomethyl)triphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
Catalog # TCIAB1206-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
(Bromomethyl)triphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
Catalog # TCIAB1206-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

About this item

 :
 :436.1200 g/mol
EINECS:213-857-2
 :GHS07
CAS Number:1034-49-7
Formula:C₁₉H₁₇Br₂P
MDL Number:MFCD00011864