Dextran sulphate sodium salt, M.W. 40000 - 50000
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9011-18-1
:
Formula:(C₆H₇Na₃O₁₄S₃)n
MDL Number:MFCD00081551
Merck Index:14,2951
Storage Temperature:Ambient
Related Information
Organic Building Blocks and Intermediates
Application Notes
Thermo Scientific Cell Culture Workflow