
2-Chloroquinoline ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:612-62-4
:
Formula:C₉H₆ClN
MDL Number:MFCD00006741
:163.61 g/mol
Boiling Pt:267°C
Melting Pt:38 °C
Density:1,2464 g/cm³ (25 °C)
Storage Temperature:Refrigerator
EINECS:210-317-8
:GHS07
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Melting point:33.0 to 38.0 deg-C
- Solubility in Methanol:almost transparency