0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
G418 disulphate
G418 disulphate
G418 disulphate
:
Cayman Chemical
G418 disulphate
:
Cayman Chemical
:
108321-42-2
:
:
C₂₀H₄₄N₄O₁₈S₂
:
MFCD00058314
:
692.72 g/mol
:
GHS07
:
Ambient