0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
1-Butyl-4-ethynylbenzene
1-Butyl-4-ethynylbenzene ≥97.0% (by GC)
1-Butyl-4-ethynylbenzene ≥97.0% (by GC)
:
TCI
1-Butyl-4-ethynylbenzene ≥97.0% (by GC)
:
TCI
:
79887-09-5
:
:
C₁₂H₁₄
:
MFCD00173885
:
158.24 g/mol
:
92°C
:
GHS07
:
Refrigerator
Appearance:
Colorless to Light orange to Yellow clear liquid
Purity (GC):
min. 97.0 %
Manual Synthetic Organic Chemistry and Materials Chemistry