Order Entry
Finland
Orders LinkContactUsLinkComponent
Benzophenone

Benzophenone

Glass bottle for solids, 100 mg
Catalog # PROOMM0630.01
Environment
Benzophenone
Catalog # PROOMM0630.01
CAS Number:  119-61-9
Benzophenone

Glass bottle for solids, 100 mg

CAS Number:  119-61-9
Price
731,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  Diphenylmethanone, Diphenyl ketone
  • CAS Number:119-61-9
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00003076
  • Molecular Weight:182.22 g/mol
  • Merck Index:13,01099
  • Boiling Pt:305.4 °C (1013 hPa)
  • Melting Pt:47…49 °C
  • Density:1.085 g/cm³ (20 °C)
  • Flash Pt:150 °C
  • Storage Temperature:Ambient
  • EINECS:204-337-6
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications