Order Entry
Finland
ContactUsLinkComponent
Plant RNA purification Mini kit, GeneJET™
Plant RNA purification Mini kit, GeneJET™
Certificates
undefined
Plant RNA purification Mini kit, GeneJET™
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Plant samples
  • Target molecule:
    RNA
  • Description:
    GeneJET™ Plant RNA Purification Mini Kit

Specifications

Related Information