VWR®, Burettes, Lateral Stopcock
Supplier: VWR Collection
Certificates
About this item
Borosilicate 3.3 glass, class AS.
- Schellbach stripe, stopcock with glass key
- High contrast blue enamel graduations
- Batch certificate included
DIN EN ISO 385
Certifications: DIN EN ISO 385