Order Entry
Finland
ContactUsLinkComponent
Surgical blades for cervical biopsy
Surgical blades for cervical biopsy
Catalog # SWAN2001
Supplier:  SWANN MORTON
undefined
Surgical blades for cervical biopsy
Catalog # SWAN2001
Supplier:  SWANN MORTON
Specifications
  • For:
    Major S/L handles
  • Description:
    Cervical biopsy blade, stainless steel, packed in individual plastic trays (10 per box)
  • Pk:
    10
Specifications
About this item
This sterile stainless cervical biposy blade is used in conjunction with the long stainless steel Major handle to take a 'cone' biopsy from the tissue in a woman's cervix. The tissue sample can then be analysed to assess the prescence of malignant and non malignant carcinoma cells.
  • Fits handles Major S and L
Handles
Product Information