Shell freezers, benchtop, FreeZone®
: LABCONCO
FreeZone® benchtop shell freezers provide a compact system for prefreezing samples for lyophilization.
- ¹/₂ hp HCFC/CFC refrigeration systems cool to −40 °C for processing samples
- LED illuminates green, when the system reaches operating temperature
- Right-side mounted power switch
- Left-side mounted retractable bath drain hose
Temperature range: − 20 °C to − 42°C
: UL certified.
- Catalog No:
- LBCP7949030
- Model:
- FreeZone®
- Cat. No.:
- LBCP7949030
- Display:
- LED
- Weight:
- 44 kg