Order Entry
Finland
ContactUsLinkComponent
Conical sleeves for vacuum filtration, Sterilin®
Conical sleeves for vacuum filtration, Sterilin®
Catalog # 511-0098
Supplier:  Sterilin
undefined
Conical sleeves for vacuum filtration, Sterilin®
Catalog # 511-0098
Supplier:  Sterilin
Specifications
  • Description:
    Set of 6 cones, assorted
  • Pk:
    1 SET
Specifications
About this item
Red natural rubber cones for use with filtering flasks.
  • Set of six pieces
  • Set includes each one of 7/15 mm, 10/22 mm, 15/28 mm, 16/36 mm, 22/42 mm and 26/52 mm (inner diamater at low end/inner diameter at top end)