Order Entry
Finland
ContactUsLinkComponent
 
Water samplers, SP 3
 :  MAXX
 
Water samplers, SP 3
 :  MAXX

 

  • Catalog No:
  • MAXA10020410
  • MAXA10020012G
  • MAXA10020012
  • MAXA1002000
  • MAXA10020210
  • MAXA10020050
  • MAXA10020406
  • MAXA10020025
  • MAXA10020414
  • Model:
  • SP III Zone 2
  • SP III Zone 2
  • SP III Zone 2
  • SP III Zone 2
  • SP III Zone 2
  • SP III Zone 2
  • SP III Zone 2
  • SP III Zone 2
  • SP III Zone 2
  • Sample size:
  • 20 − 350 ml
  • 20 − 350 ml
  • 20 − 350 ml
  • 20 − 350 ml
  • 20 − 350 ml
  • 20 − 350 ml
  • 20 − 350 ml
  • 20 − 350 ml
  • 20 − 350 ml
  • Programs:
  • Six user-defined programs with start, pause and stop options and pulse settings
  • Six user-defined programs with start, pause and stop options and pulse settings
  • Six user-defined programs with start, pause and stop options and pulse settings
  • Six user-defined programs with start, pause and stop options and pulse settings
  • Six user-defined programs with start, pause and stop options and pulse settings
  • Six user-defined programs with start, pause and stop options and pulse settings
  • Six user-defined programs with start, pause and stop options and pulse settings
  • Six user-defined programs with start, pause and stop options and pulse settings
  • Six user-defined programs with start, pause and stop options and pulse settings
  • Datalog:
  • Non volatile data memory for storing data/messages
  • Non volatile data memory for storing data/messages
  • Non volatile data memory for storing data/messages
  • Non volatile data memory for storing data/messages
  • Non volatile data memory for storing data/messages
  • Non volatile data memory for storing data/messages
  • Non volatile data memory for storing data/messages
  • Non volatile data memory for storing data/messages
  • Non volatile data memory for storing data/messages
  • Accuracy:
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • 2,8 %
  • Signal input:
  • 1× analogue ; 5× digital
  • 1× analogue ; 5× digital
  • 1× analogue ; 5× digital
  • 1× analogue ; 5× digital
  • 1× analogue ; 5× digital
  • 1× analogue ; 5× digital
  • 1× analogue ; 5× digital
  • 1× analogue ; 5× digital
  • 1× analogue ; 5× digital
  • Signal output:
  • Up to 8× digital
  • Up to 8× digital
  • Up to 8× digital
  • Up to 8× digital
  • Up to 8× digital
  • Up to 8× digital
  • Up to 8× digital
  • Up to 8× digital
  • Up to 8× digital
  • Communication interface:
  • GSM (optional)
  • GSM (optional)
  • GSM (optional)
  • GSM (optional)
  • GSM (optional)
  • GSM (optional)
  • GSM (optional)
  • GSM (optional)
  • GSM (optional)
  • Housing:
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Control:
  • Microprocessor with 4 MB Flashrom, 32 KB RAM (battery buffered), 32 KB EEprom and real-time clock
  • Microprocessor with 4 MB Flashrom, 32 KB RAM (battery buffered), 32 KB EEprom and real-time clock
  • Microprocessor with 4 MB Flashrom, 32 KB RAM (battery buffered), 32 KB EEprom and real-time clock
  • Microprocessor with 4 MB Flashrom, 32 KB RAM (battery buffered), 32 KB EEprom and real-time clock
  • Microprocessor with 4 MB Flashrom, 32 KB RAM (battery buffered), 32 KB EEprom and real-time clock
  • Microprocessor with 4 MB Flashrom, 32 KB RAM (battery buffered), 32 KB EEprom and real-time clock
  • Microprocessor with 4 MB Flashrom, 32 KB RAM (battery buffered), 32 KB EEprom and real-time clock
  • Microprocessor with 4 MB Flashrom, 32 KB RAM (battery buffered), 32 KB EEprom and real-time clock
  • Microprocessor with 4 MB Flashrom, 32 KB RAM (battery buffered), 32 KB EEprom and real-time clock
  • Suction height:
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • <8 mWC
  • Power requirements:
  • 350 V
  • 350 V
  • 350 V
  • 350 V
  • 350 V
  • 350 V
  • 350 V
  • 350 V
  • 350 V
  • W×D×H:
  • 690×645×1470 mm
  • 690×645×1470 mm
  • 690×645×1470 mm
  • 690×645×1470 mm
  • 690×645×1470 mm
  • 690×645×1470 mm
  • 690×645×1470 mm
  • 690×645×1470 mm
  • 690×645×1470 mm
  • Weight:
  • 100 kg
  • 100 kg
  • 100 kg
  • 100 kg
  • 100 kg
  • 100 kg
  • 100 kg
  • 100 kg
  • 100 kg