Transfer pipettes, Sterilin®
Supplier: Sterilin
Certificates
About this item
These disposable pipettes, manufactured from non toxic LDPE, transfer liquids safely and accurately. Ideal for general laboratory use, sample storage and transport.
- Integral filling bulb eliminates contamination associated with separate rubber bulbs
- Excellent transparency and uniform thickness
- Can be heat sealed for sample storage and transportation
- Heat-sealed bulbs suitable for storage in the gas phase of liquid nitrogen
- 1 ml graduated version: Suitable for general laboratory use, storage and transportation of samples
- 3 ml graduated version: Suitable for blood banking and general laboratory use
- Narrow stem version: Suitable for use as a freeze vial, sedimentation and transport