

Acridine orange (hemizinc salt) 85% (dye content), Sigma-Aldrich®
:10127-02-3
:
:C₁₇H₂₀ClN₃·HCl·½ZnCl₂
:MFCD00081043
:369.96 g/mol
:M-Clarity™ quality level = MQ200
:233-353-6
:GHS08
:GHS07
:Room Temperature
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Wavelength:489 - 495 nm
- Extinction Coefficient:≥38000
- Carbon:≥46.3%
- Nitrogen:≥9.5%
- ICP Major Analysis Zinc Component:Confirmed
- Solubility (Turbidity):Clear
- Solubility (Colour):Orange to very dark orange and brown to very dark brown and red-orange and red-brown and orange-brown and very dark red-brown