

Acridine orange (hemizinc salt)
Catalog # MOLEM17692553
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:369.96 g/mol
EINECS:233-353-6
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:10127-02-3
Formula:C₁₇H₂₀ClN₃·HCl·½ZnCl₂
MDL Number:MFCD00081043