Order Entry
Finland
ContactUsLinkComponent
Acridine orange (chlorid salt) 10 mg/ml in water, high purity (AO)
Health
Acridine orange (chlorid salt) 10 mg/ml in water, high purity (AO)
Catalog # 40039.
Supplier:  Biotium
CAS Number:  
undefined
Health
Acridine orange (chlorid salt) 10 mg/ml in water, high purity (AO)
Catalog # 40039.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :301.81 g/mol (anhy)
EINECS:233-353-6
 :GHS08
CAS Number:65-61-2
 :1704465-79-1
Formula:C₁₇H₁₉N₃·HCl·xH₂O
MDL Number:MFCD00012660