Order Entry
Finland
ContactUsLinkComponent
2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers) 97% (mixture of cis and trans)
Flammable
Irritant
2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers) 97% (mixture of cis and trans)
Catalog # ACRO115531000
CAS Number:  
undefined
Flammable
Irritant
2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers) 97% (mixture of cis and trans)
Catalog # ACRO115531000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4330 to 1.4350 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :130.14 g/mol
Boiling Pt:160…162 °C (760 mmHg)
Density:1.07 g/cm³
Flash Pt:47 °C
Storage Temperature:Ambient
EINECS:206-367-5
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:332-77-4
Formula:C₆H₁₀O₃
MDL Number:MFCD00003220