Order Entry
Finland
ContactUsLinkComponent
Acriflavine
Acriflavine
Catalog # MOLEM78060746
Supplier:  Molekula
CAS Number:  
Acriflavine
Catalog # MOLEM78060746
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :468.9806 g/mol
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:8048-52-0
 :65589-70-0
Formula:C₂₇H₂₅ClN₆
MDL Number:MFCD00064307