Order Entry
Finland
ContactUsLinkComponent
Guanosine 5'-triphosphate disodium salt, Millipore®
Guanosine 5'-triphosphate disodium salt, Millipore®
Catalog # 371701-250
CAS Number:  
Guanosine 5'-triphosphate disodium salt, Millipore®
Catalog # 371701-250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :567.147 g/mol
EINECS:259-940-7
CAS Number:56001-37-7
 :MFCD00083629
Formula:C₁₀H₁₄N₅Na₂O₁₄P₃
MDL Number:MFCD03410297