Ponceau S, Bioreagent for electrophoresis, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:6226-79-5
:
Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number:MFCD00003892
:760.56 g/mol
:M-Clarity™ quality level = MQ200
EINECS:228-319-2
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Infrared spectrum:Conforms to Structure
- Solubility (Turbidity):Clear
- Solubility (Colour)
c = 4mg/4ml; Water:Red to Very Dark Red - Suitability
Suitable for Use as a Stain in Electrophoresis:Suitable - Wavelength
C = 0.015 G/L, Di water:518 - 522 nm - Extinction Coefficient:≥33000