Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ponceau S
Ponceau S
Ponceau S
Ponceau S
MOLE43797916-25G
:
Molekula
:
Ponceau S
MOLE43797916-25G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
760.56 g/mol
:
228-319-2
:
GHS07
:
Ambient
:
6226-79-5
:
C₂₂H₁₂N₄Na₄O₁₃S₄
:
MFCD00003892