Order Entry
Finland
ContactUsLinkComponent
Azure A, Sigma-Aldrich®
Azure A, Sigma-Aldrich®
Catalog # A2918-25G
CAS Number:  
undefined
Azure A, Sigma-Aldrich®
Catalog # A2918-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :291.797 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:208-510-7
 :GHS07
Storage Temperature:Ambient
CAS Number:531-53-3
Formula:C₁₄H₁₄ClN₃S
MDL Number:MFCD00012112