
Alizarin red S (sodium salt)
Supplier: Merck
Certificates
About this item
CAS Number:130-22-3
:93982-72-0
Formula:C₁₄H₇NaO₇S
MDL Number:MFCD00013049
:342.26 g/mol
:M-Clarity™ quality level = MQ200
EINECS:204-981-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:342.26 g/mol